3-Bromo-5-formyl-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

69356-500MG 69356-1G 69356-5G
77337-222EA 408.91 USD
77337-222 77357-650 77381-596
3-Bromo-5-formyl-2-methoxybenzoic acid ≥97%
3-Bromo-5-formyl-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇BrO₄
Storage Temperature: Ambient
MDL Number: MFCD34181063
CAS Number: 2248321-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR