3-Bromo-5-formyl-2-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

69352-500MG 69352-1G 69352-5G 69352-25G
77333-510EA 265.74 USD
77333-510 77357-648 77369-518 77390-500
3-Bromo-5-formyl-2-isopropoxybenzoic acid ≥97%
3-Bromo-5-formyl-2-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₄
Storage Temperature: Ambient
MDL Number: MFCD34763908
CAS Number: 2807470-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR