5-Fluoro-2,3'-dimethyl-[1,1'-biphenyl]-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

69349-500MG 69349-1G 69349-5G
77336-894EA 396.55 USD
77336-894 77357-646 77381-594
5-Fluoro-2,3'-dimethyl-[1,1'-biphenyl]-4-carboxylic acid ≥97%
5-Fluoro-2,3'-dimethyl-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
MW: 244.261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763907
CAS Number: 2901099-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR