2-Bromo-3,4-dichloro-1-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

69335-500MG 69335-1G 69335-5G 69335-25G
77333-506EA 134.93 USD
77333-506 77344-488 77369-516 77390-498
2-Bromo-3,4-dichloro-1-methoxybenzene ≥95%
2-Bromo-3,4-dichloro-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
MW: 255.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31725577
CAS Number: 174913-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR