(2,3-Dichloro-6-methoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

69334-500MG 69334-1G 69334-5G 69334-25G
77333-504EA 114.33 USD
77333-504 77343-372 77369-514 77381-586
(2,3-Dichloro-6-methoxyphenyl)(methyl)sulfane ≥95%
(2,3-Dichloro-6-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈Cl₂OS
Storage Temperature: Ambient
MDL Number: MFCD34763897
CAS Number: 2705865-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR