(3-Bromo-5-chloro-2-fluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

69324-500MG 69324-1G 69324-5G 69324-25G
77333-502EA 204.97 USD
77333-502 77357-638 77369-512 77390-496
(3-Bromo-5-chloro-2-fluorophenyl)trimethylsilane ≥95%
(3-Bromo-5-chloro-2-fluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₁BrClFSi
Storage Temperature: Ambient
MDL Number: MFCD34763887
CAS Number: 2901082-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR