(5-Chloro-2-fluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

69321-500MG 69321-1G 69321-5G 69321-25G
77333-500EA 157.59 USD
77333-500 77345-668 77369-510 77390-494
(5-Chloro-2-fluorophenyl)trimethylsilane ≥97%
(5-Chloro-2-fluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₂ClFSi
Storage Temperature: Ambient
MDL Number: MFCD30178314
CAS Number: 2095209-09-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR