2-Ethyl-1-fluoro-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

69315-500MG 69315-1G 69315-5G
77339-562EA 596.37 USD
77339-562 77357-636 77381-584
2-Ethyl-1-fluoro-4-methylbenzene ≥95%
2-Ethyl-1-fluoro-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁F
MW: 138.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22489445
CAS Number: 1369903-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR