(2,5-Difluoro-1,4-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

69300-1G 69300-5G 69300-25G
77341-838EA 105.06 USD
77341-838 77369-506 77381-582
(2,5-Difluoro-1,4-phenylene)bis(methylsulfane) ≥97%
(2,5-Difluoro-1,4-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₈H₈F₂S₂
MW: 206.276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763879
CAS Number: 2807467-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR