(2,5-Difluoro-4-iodophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

69299-500MG 69299-1G 69299-5G 69299-25G
77333-496EA 99.91 USD
77333-496 77342-880 77369-504 77381-580
(2,5-Difluoro-4-iodophenyl)(methyl)sulfane ≥95%
(2,5-Difluoro-4-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅F₂IS
MW: 286.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763878
CAS Number: 2706161-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR