2-Bromo-5-isobutoxy-1,3-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

69286-500MG 69286-1G 69286-5G 69286-25G
77333-494EA 120.51 USD
77333-494 77343-808 77369-502 77381-578
2-Bromo-5-isobutoxy-1,3-dimethylbenzene ≥95%
2-Bromo-5-isobutoxy-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₇BrO
MW: 257.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917834
CAS Number: 91560-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR