(4-Butoxy-2,6-dimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

69285-500MG 69285-1G 69285-5G 69285-25G
77333-492EA 186.43 USD
77333-492 77357-632 77369-500 77390-490
(4-Butoxy-2,6-dimethylphenyl)(methyl)sulfane ≥97%
(4-Butoxy-2,6-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₃H₂₀OS
Storage Temperature: Ambient
MDL Number: MFCD34763871
CAS Number: 2901082-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR