5-Butoxy-2-iodo-1,3-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

69284-500MG 69284-1G 69284-5G 69284-25G
77333-490EA 154.5 USD
77333-490 77345-580 77369-498 77390-488
5-Butoxy-2-iodo-1,3-dimethylbenzene ≥95%
5-Butoxy-2-iodo-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₇IO
MW: 304.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763870
CAS Number: 2901097-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR