2-Benzyl-4-bromo-1-isopropoxybenzene ≥95%

Supplier: AOBChem USA

Warning

69266-500MG 69266-1G 69266-5G
77339-102EA 550.02 USD
77339-102 77357-626 77381-574
2-Benzyl-4-bromo-1-isopropoxybenzene ≥95%
2-Benzyl-4-bromo-1-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₁₆H₁₇BrO
MW: 305.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763862
CAS Number: 2901083-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR