2-Benzyl-4-bromo-1-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

69265-500MG 69265-1G 69265-5G
77333-482EA 309 USD
77333-482 77357-624 77381-572
2-Benzyl-4-bromo-1-ethoxybenzene ≥97%
2-Benzyl-4-bromo-1-ethoxybenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO
MW: 291.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763861
CAS Number: 2901085-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR