N-([1,1'-biphenyl]-2-yl)acetamide ≥97%

Supplier: AOBChem USA

Warning

69259-500MG 69259-1G 69259-5G 69259-25G
77333-480EA 282.22 USD
77333-480 77357-622 77369-492 77390-482
N-([1,1'-biphenyl]-2-yl)acetamide ≥97%
N-([1,1'-biphenyl]-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD00092355
CAS Number: 2113-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR