1-Bromo-3-fluoro-2-iodo-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

69258-500MG 69258-1G 69258-5G 69258-25G
77333-478EA 178.19 USD
77333-478 77346-692 77369-490 77390-480
1-Bromo-3-fluoro-2-iodo-4-methylbenzene ≥97%
1-Bromo-3-fluoro-2-iodo-4-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
MW: 314.921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28739622
CAS Number: 1805420-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR