4-Bromo-2-methoxy-4'-(trifluoromethoxy)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

69257-500MG 69257-1G 69257-5G 69257-25G
77333-476EA 141.11 USD
77333-476 77344-906 77369-488 77390-478
4-Bromo-2-methoxy-4'-(trifluoromethoxy)-1,1'-biphenyl ≥95%
4-Bromo-2-methoxy-4'-(trifluoromethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34763858
CAS Number: 2901102-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR