6-Bromo-2-fluoro-3-(methylsulfonyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

69252-500MG 69252-1G 69252-5G 69252-25G
77333-474EA 122.57 USD
77333-474 77344-008 77369-486 77381-570
6-Bromo-2-fluoro-3-(methylsulfonyl)pyridine ≥97%
6-Bromo-2-fluoro-3-(methylsulfonyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNO₂S
MW: 254.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763856
CAS Number: 2807465-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR