2,5-Dibromo-1-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

69239-500MG 69239-1G 69239-5G
77333-472EA 322.39 USD
77333-472 77357-620 77381-568
2,5-Dibromo-1-iodo-3-methylbenzene ≥97%
2,5-Dibromo-1-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Br₂I
MW: 375.827 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763852
CAS Number: 2386569-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR