2,6-Dichloro-4-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

69233-500MG 69233-1G 69233-5G 69233-25G
77333-468EA 153.47 USD
77333-468 77345-522 77369-482 77390-474
2,6-Dichloro-4-ethoxybenzoic acid ≥97%
2,6-Dichloro-4-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₃
MW: 235.064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763849
CAS Number: 2386922-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR