(2,6-Dichloro-4-ethoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

69232-500MG 69232-1G 69232-5G 69232-25G
77333-466EA 141.11 USD
77333-466 77344-904 77369-480 77390-472
(2,6-Dichloro-4-ethoxyphenyl)(methyl)sulfane ≥97%
(2,6-Dichloro-4-ethoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀Cl₂OS
MW: 237.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763848
CAS Number: 2710538-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR