(2,6-Dichloro-4-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

69231-500MG 69231-1G 69231-5G 69231-25G
77333-464EA 260.59 USD
77333-464 77357-616 77369-478 77390-470
(2,6-Dichloro-4-methoxyphenyl)(methyl)sulfane ≥97%
(2,6-Dichloro-4-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈Cl₂OS
MW: 223.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763847
CAS Number: 2711335-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR