2-Fluoro-3,4-bis(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

69226-500MG 69226-1G 69226-5G
77333-462EA 322.39 USD
77333-462 77357-614 77381-566
2-Fluoro-3,4-bis(methylthio)pyridine ≥97%
2-Fluoro-3,4-bis(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈FNS₂
MW: 189.274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763845
CAS Number: 2807447-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR