3-Bromo-2-fluoro-4-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

69225-500MG 69225-1G 69225-5G
77335-910EA 367.71 USD
77335-910 77357-612 77381-564
3-Bromo-2-fluoro-4-(methylthio)pyridine ≥97%
3-Bromo-2-fluoro-4-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNS
MW: 222.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763844
CAS Number: 2901065-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR