(5-Bromo-2-fluoro-3-methoxyphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

69197-1G 69197-5G 69197-25G
77341-438EA 85.49 USD
77341-438 77369-476 77381-556
(5-Bromo-2-fluoro-3-methoxyphenyl)trimethylsilane ≥97%
(5-Bromo-2-fluoro-3-methoxyphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrFOSi
MW: 277.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763832
CAS Number: 2807454-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR