2-Bromo-4'-methoxy-4-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

69191-500MG 69191-1G 69191-5G 69191-25G
77333-458EA 184.37 USD
77333-458 77347-326 77369-474 77390-468
2-Bromo-4'-methoxy-4-methyl-1,1'-biphenyl ≥95%
2-Bromo-4'-methoxy-4-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
MW: 277.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763827
CAS Number: 2807470-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR