2-Bromo-2',4-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

69190-500MG 69190-1G 69190-5G 69190-25G
77333-456EA 120.51 USD
77333-456 77343-806 77369-472 77381-554
2-Bromo-2',4-dimethyl-1,1'-biphenyl ≥97%
2-Bromo-2',4-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃Br
Storage Temperature: Ambient
MDL Number: MFCD34763826
CAS Number: 1171900-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR