2-Bromo-3',4-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

69189-500MG 69189-1G 69189-5G 69189-25G
77333-454EA 120.51 USD
77333-454 77343-804 77369-470 77381-552
2-Bromo-3',4-dimethyl-1,1'-biphenyl ≥97%
2-Bromo-3',4-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃Br
MW: 261.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763825
CAS Number: 2807465-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR