2-Bromo-5-methoxy-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69183-500MG 69183-1G 69183-5G 69183-25G
77333-452EA 141.11 USD
77333-452 77344-902 77369-468 77390-466
2-Bromo-5-methoxy-4-methylbenzaldehyde ≥97%
2-Bromo-5-methoxy-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977474
CAS Number: 57295-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR