4,6-Difluoroisophthalaldehyde ≥97%

Supplier: AOBChem USA

Warning

69180-500MG 69180-1G 69180-5G 69180-25G
77333-450EA 135.96 USD
77333-450 77344-518 77369-466 77390-464
4,6-Difluoroisophthalaldehyde ≥97%
4,6-Difluoroisophthalaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22042483
CAS Number: 397299-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR