2,4-Difluoro-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69179-500MG 69179-1G 69179-5G 69179-25G
77333-448EA 182.31 USD
77333-448 77347-014 77369-464 77390-462
2,4-Difluoro-5-(methylthio)benzaldehyde ≥97%
2,4-Difluoro-5-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂OS
MW: 188.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763823
CAS Number: 2901102-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR