2,4-Difluoro-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

69177-500MG 69177-1G 69177-5G
77338-852EA 525.3 USD
77338-852 77357-604 77381-550
2,4-Difluoro-5-formylbenzoic acid ≥97%
2,4-Difluoro-5-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₂O₃
MW: 186.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12196969
CAS Number: 1162674-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR