3,4-Dichloro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69174-500MG 69174-1G 69174-25G 69174-5G
77333-446EA 285.31 USD
77333-446 77357-602 77390-460 77381-548
3,4-Dichloro-5-methylbenzaldehyde ≥97%
3,4-Dichloro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD24682344
CAS Number: 170879-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR