(3-Bromo-5-methylphenoxy)(tert-butyl)dimethylsilane ≥95%

Supplier: AOBChem USA

Warning

69167-500MG 69167-1G 69167-5G 69167-25G
77333-444EA 116.39 USD
77333-444 77343-458 77369-460 77381-544
(3-Bromo-5-methylphenoxy)(tert-butyl)dimethylsilane ≥95%
(3-Bromo-5-methylphenoxy)(tert-butyl)dimethylsilane

Precaución: for Research Use Only

Formula: C₁₃H₂₁BrOSi
MW: 301.295 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30296253
CAS Number: 136386-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR