1-Bromo-3-isobutoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

69166-500MG 69166-1G 69166-5G 69166-25G
77333-442EA 134.93 USD
77333-442 77344-502 77369-458 77390-458
1-Bromo-3-isobutoxy-5-methylbenzene ≥97%
1-Bromo-3-isobutoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043850
CAS Number: 1881330-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR