3-(Difluoromethoxy)-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69164-500MG 69164-1G 69164-5G
77340-186EA 820.91 USD
77340-186 77357-600 77381-542
3-(Difluoromethoxy)-5-methylbenzaldehyde ≥97%
3-(Difluoromethoxy)-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34763818
CAS Number: 2384756-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR