1-Bromo-2-ethoxy-3,4-difluoro-5-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

69150-500MG 69150-1G 69150-25G 69150-5G
77333-438EA 285.31 USD
77333-438 77357-598 77390-456 77381-538
1-Bromo-2-ethoxy-3,4-difluoro-5-(methylsulfonyl)benzene ≥97%
1-Bromo-2-ethoxy-3,4-difluoro-5-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₉H₉BrF₂O₃S
MW: 315.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763809
CAS Number: 2807470-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR