2-Ethoxy-3,4-difluoro-5-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

69147-500MG 69147-1G 69147-5G
77339-452EA 590.19 USD
77339-452 77357-596 77381-536
2-Ethoxy-3,4-difluoro-5-methylbenzoic acid ≥95%
2-Ethoxy-3,4-difluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
MW: 216.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763806
CAS Number: 2807464-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR