(4'-Isopropoxy-[1,1'-biphenyl]-4-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

69142-500MG 69142-1G 69142-5G 69142-25G
77333-432EA 99.91 USD
77333-432 77342-878 77369-450 77381-532
(4'-Isopropoxy-[1,1'-biphenyl]-4-yl)(methyl)sulfane ≥97%
(4'-Isopropoxy-[1,1'-biphenyl]-4-yl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₆H₁₈OS
MW: 258.379 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763801
CAS Number: 2807450-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR