1-(5-Propylthiophen-2-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

69132-1G 69132-5G 69132-25G
77340-910EA 62.83 USD
77340-910 77369-448 77381-530
1-(5-Propylthiophen-2-yl)ethanone ≥97%
1-(5-Propylthiophen-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₉H₁₂OS
MW: 168.256 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04967420
CAS Number: 832737-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR