5-Acetyl-2-chlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69113-500MG 69113-1G 69113-5G
77337-220EA 408.91 USD
77337-220 77357-588 77381-526
5-Acetyl-2-chlorobenzaldehyde ≥97%
5-Acetyl-2-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇ClO₂
Storage Temperature: Ambient
MDL Number: MFCD26679282
CAS Number: 1500017-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR