5-Bromo-4-chloro-2-fluoro-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

69107-500MG 69107-1G 69107-5G 69107-25G
77333-426EA 165.83 USD
77333-426 77346-176 77369-442 77390-448
5-Bromo-4-chloro-2-fluoro-3-formylbenzoic acid ≥97%
5-Bromo-4-chloro-2-fluoro-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₃BrClFO₃
Storage Temperature: Ambient
CAS Number: 2918958-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR