3-Ethoxy-4-fluoro-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

69098-500MG 69098-1G
77340-478EA 1221.58 USD
77340-478 77357-584
3-Ethoxy-4-fluoro-5-formylbenzoic acid ≥97%
3-Ethoxy-4-fluoro-5-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉FO₄
MW: 212.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763783
CAS Number: 2386837-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR