5-Bromo-1-ethoxy-2-fluoro-3-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

69097-500MG 69097-1G 69097-5G 69097-25G
77333-424EA 214.24 USD
77333-424 77357-582 77369-440 77390-446
5-Bromo-1-ethoxy-2-fluoro-3-(methoxymethyl)benzene ≥97%
5-Bromo-1-ethoxy-2-fluoro-3-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO₂
MW: 263.103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763782
CAS Number: 2901082-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR