4-Fluoro-3-formyl-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

69095-500MG 69095-1G 69095-5G
77340-402EA 1017.64 USD
77340-402 77357-578 77381-522
4-Fluoro-3-formyl-5-methoxybenzoic acid ≥97%
4-Fluoro-3-formyl-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₄
Storage Temperature: Ambient
MDL Number: MFCD34763780
CAS Number: 2386117-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR