5-Bromo-4'-(trifluoromethoxy)-[1,1'-biphenyl]-2-carbonitrile ≥97%

Supplier: AOBChem USA

Warning

69094-500MG 69094-1G 69094-5G 69094-25G
77333-422EA 184.37 USD
77333-422 77347-324 77369-438 77390-444
5-Bromo-4'-(trifluoromethoxy)-[1,1'-biphenyl]-2-carbonitrile ≥97%
5-Bromo-4'-(trifluoromethoxy)-[1,1'-biphenyl]-2-carbonitrile
Formula: C₁₄H₇BrF₃NO
Storage Temperature: Ambient
MDL Number: MFCD34763779
CAS Number: 2901098-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR