5-Bromo-4'-formyl-[1,1'-biphenyl]-2-carbonitrile ≥95%

Supplier: AOBChem USA

Warning

69091-500MG 69091-1G 69091-5G 69091-25G
77333-418EA 184.37 USD
77333-418 77347-322 77369-434 77390-440
5-Bromo-4'-formyl-[1,1'-biphenyl]-2-carbonitrile ≥95%
5-Bromo-4'-formyl-[1,1'-biphenyl]-2-carbonitrile

Precaución: for Research Use Only

Formula: C₁₄H₈BrNO
MW: 286.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763777
CAS Number: 2901083-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR