2'-Fluoro-3'-methyl-[1,1'-biphenyl]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

69083-500MG 69083-1G 69083-5G 69083-25G
77333-416EA 202.91 USD
77333-416 77357-572 77369-432 77390-438
2'-Fluoro-3'-methyl-[1,1'-biphenyl]-3-carbaldehyde ≥97%
2'-Fluoro-3'-methyl-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO
MW: 214.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26721635
CAS Number: 1522512-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR