2,4'-Difluoro-3-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

69078-500MG 69078-1G 69078-5G
77340-328EA 900.22 USD
77340-328 77357-568 77381-518
2,4'-Difluoro-3-methyl-1,1'-biphenyl ≥97%
2,4'-Difluoro-3-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₂
MW: 204.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14700883
CAS Number: 1214367-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR